Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Edem1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_238366.8 | XP_238366.4 | 4 - 5 | 961 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Rattus norvegicus Probe spans exons |