Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Iqsec1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006236890.2 | XP_006236952.1 | 10 - 11 | 3058 | 67 | |
XM_008763140.1 | XP_008761362.1 | 10 - 11 | 3058 | 67 | ||
XM_008763141.1 | XP_008761363.1 | 10 - 11 | 3058 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Rattus norvegicus Probe spans exons |