Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Iqsec1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006236890.2 | XP_006236952.1 | 13 - 14 | 3453 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Rattus norvegicus Probe spans exons |