Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dcun1d4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108359.1 | NP_001101829.1 | 7 - 8 | 587 | 139 | |
XM_006250907.2 | XP_006250969.1 | 5 - 6 | 486 | 139 | ||
XM_008770158.1 | XP_008768380.1 | 7 - 8 | 598 | 139 | ||
XM_008770159.1 | XP_008768381.1 | 5 - 6 | 389 | 139 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Rattus norvegicus Probe spans exons |