Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gabrg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_080586.1 | NP_542153.1 | 4 - 5 | 638 | 76 | |
GenBank mRNA | X57514.1 | - | 4 - 5 | 638 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Ion Channel Rattus norvegicus Probe spans exons |