Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Katna1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004217.4 | NP_001004217.2 | 9 - 10 | 1345 | 92 | |
GenBank mRNA | AY621629.1 | - | 8 - 9 | 1149 | 92 | |
BC097929.1 | - | 9 - 10 | 1345 | 7444887 | 92 | |
BI302698.1 | - | 2 - 3 | 92 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |