Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il22ra2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003404.1 | NP_001003404.1 | 2 - 3 | 224 | 111 | |
XM_006227664.2 | XP_006227726.1 | 3 - 4 | 277 | 111 | ||
GenBank mRNA | AJ555485.1 | - | 2 - 3 | 246 | 111 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |