Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dpcd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013905.1 | NP_001013927.1 | 1 - 2 | 95 | 114 | |
XM_006231450.2 | XP_006231512.1 | 1 - 2 | 125 | 114 | ||
GenBank mRNA | BC078988.1 | - | 1 - 2 | 95 | 7112530 | 114 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |