Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dlat | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031025.1 | NP_112287.1 | 6 - 7 | 957 | 135 | |
GenBank mRNA | BC107440.1 | - | 6 - 7 | 957 | 7115402 | 135 |
D00092.1 | - | 3 - 4 | 461 | 135 | ||
D10655.1 | - | 5 - 6 | 674 | 135 | ||
M16075.1 | - | 3 - 4 | 461 | 135 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |