Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rdx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005889.2 | NP_001005889.2 | 2 - 3 | 219 | 141 | |
GenBank mRNA | BC087147.1 | - | 2 - 3 | 219 | 7130553 | 141 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |