Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rdx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005889.2 | NP_001005889.2 | 4 - 5 | 408 | 164 | |
GenBank mRNA | AY428868.1 | - | 3 - 4 | 233 | 164 | |
BC087147.1 | - | 4 - 5 | 408 | 7130553 | 164 | |
KF307742.1 | - | 3 - 4 | 251 | 164 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |