Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hmg20a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108150.1 | NP_001101620.1 | 2 - 3 | 365 | 64 | |
XM_006243117.2 | XP_006243179.1 | 2 - 3 | 3922 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus Probe spans exons |