Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pml | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008766431.1 | XP_008764653.1 | 5 - 6 | 1653 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Rattus norvegicus Probe spans exons |