Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cep57l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017448.1 | NP_001017448.1 | 2 - 3 | 258 | 77 | |
XM_006256562.2 | XP_006256624.1 | 2 - 3 | 539 | 77 | ||
XM_006256563.2 | XP_006256625.1 | 2 - 3 | 542 | 77 | ||
XM_006256564.2 | XP_006256626.1 | 2 - 3 | 294 | 77 | ||
XM_006256565.1 | XP_006256627.1 | 3 - 4 | 317 | 77 | ||
XM_006256566.2 | XP_006256628.1 | 2 - 3 | 951 | 77 | ||
XM_006256567.2 | XP_006256629.1 | 3 - 4 | 286 | 77 | ||
XM_006256568.2 | XP_006256630.1 | 1 - 2 | 169 | 77 | ||
XM_008772982.1 | XP_008771204.1 | 2 - 3 | 1209 | 77 | ||
XM_008772983.1 | XP_008771205.1 | 2 - 3 | 282 | 77 | ||
GenBank mRNA | BC079256.1 | - | 2 - 3 | 258 | 7133287 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |