Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rtn4ip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107644.3 | NP_001101114.1 | 7 - 8 | 1389 | 71 | |
XM_008772999.1 | XP_008771221.1 | 8 - 9 | 1078 | 71 | ||
GenBank mRNA | BC082054.1 | - | 8 - 9 | 1731 | 71 | |
BC166465.1 | - | 7 - 8 | 1393 | 9030981 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |