Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006242814.2 | XP_006242876.1 | 10 - 11 | 1940 | 111 | |
XM_006242815.2 | XP_006242877.2 | 10 - 11 | 1929 | 111 | ||
XM_008766078.1 | XP_008764300.1 | 10 - 11 | 1929 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |