Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Galntl6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001135756.1 | NP_001129228.1 | 10 - 11 | 1642 | 66 | |
XM_008771227.1 | XP_008769449.1 | 9 - 10 | 2296 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Rattus norvegicus Probe spans exons |