Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Alg12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108104.2 | NP_001101574.2 | 4 - 5 | 514 | 82 | |
GenBank mRNA | BC161934.1 | - | 4 - 5 | 512 | 9022144 | 82 |
FM050024.1 | - | 4 - 5 | 514 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |