Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kdelr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001127546.1 | NP_001121018.1 | 2 - 3 | 263 | 108 | |
XM_008765714.1 | XP_008763936.1 | 2 - 3 | 981 | 108 | ||
GenBank mRNA | BC162033.1 | - | 2 - 3 | 263 | 9103157 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |