Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Naga | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012120.1 | NP_001012120.1 | 1 - 2 | 63 | 59 | |
XM_006242087.2 | XP_006242149.1 | 2 - 3 | 277 | 59 | ||
XM_006242088.2 | XP_006242150.1 | 2 - 3 | 213 | 59 | ||
GenBank mRNA | BC082084.1 | - | 1 - 2 | 63 | 7133636 | 59 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Rattus norvegicus Probe spans exons |