Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Shmt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008322.1 | NP_001008323.1 | 8 - 9 | 1082 | 71 | |
XM_006241453.2 | XP_006241515.1 | 8 - 9 | 1108 | 71 | ||
GenBank mRNA | BC085331.1 | - | 8 - 9 | 1082 | 7106911 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |