Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem71 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271215.1 | NP_001258144.1 | 2 | 245 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Rattus norvegicus Both primers and probe map within a single exon |