Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Invs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107932.1 | NP_001101402.1 | 5 - 6 | 976 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |