Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Naaa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001010967.2 | NP_001010967.1 | 3 - 4 | 528 | 74 | |
GenBank mRNA | AB162193.1 | - | 3 - 4 | 526 | 74 | |
BC105771.1 | - | 3 - 4 | 528 | 7930474 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |