Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pknox1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013074.1 | NP_001013092.1 | 8 - 9 | 1001 | 72 | |
XM_006256187.2 | XP_006256249.1 | 9 - 10 | 2036 | 72 | ||
XM_006256188.2 | XP_006256250.1 | 10 - 11 | 2098 | 72 | ||
XM_006256189.2 | XP_006256251.1 | 10 - 11 | 2090 | 72 | ||
XM_006256190.2 | XP_006256252.1 | 9 - 10 | 2034 | 72 | ||
XM_008772808.1 | XP_008771030.1 | 10 - 11 | 1913 | 72 | ||
XM_008772809.1 | XP_008771031.1 | 9 - 10 | 2028 | 72 | ||
XM_008772810.1 | XP_008771032.1 | 9 - 10 | 1916 | 72 | ||
GenBank mRNA | BC091397.1 | - | 8 - 9 | 1001 | 7323315 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |