Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Srpk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025726.1 | NP_001020897.1 | 14 - 15 | 1764 | 83 | |
XM_008772734.1 | XP_008770956.1 | 14 - 15 | 1885 | 83 | ||
GenBank mRNA | BC099089.1 | - | 14 - 15 | 1764 | 7456350 | 83 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protein Kinase Mammalian Gene Collection Rattus norvegicus Probe spans exons |