Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fgd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107617.2 | NP_001101087.2 | 4 - 5 | 709 | 134 | |
XM_006256205.2 | XP_006256267.1 | 4 - 5 | 755 | 134 | ||
XM_008772729.1 | XP_008770951.1 | 4 - 5 | 755 | 134 | ||
GenBank mRNA | BC089841.1 | - | 4 - 5 | 681 | 134 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |