Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Alk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001169101.2 | NP_001162572.1 | 4105 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Protein Kinase Rattus norvegicus Probe spans exons |