Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Six6os1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006240218.2 | XP_006240280.1 | 11 - 12 | 1015 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |