Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Morn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001126086.2 | NP_001119558.1 | 3 - 4 | 285 | 82 | |
NM_001145449.1 | NP_001138921.1 | 3 - 4 | 260 | 82 | ||
XM_006239651.2 | XP_006239713.2 | 3 - 4 | 1340 | 82 | ||
GenBank mRNA | BC166893.1 | - | 3 - 4 | 264 | 6919817 | 82 |
CB765303.1 | - | 3 - 4 | 260 | 82 | ||
FM039224.1 | - | 3 - 4 | 248 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |