Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Morn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001126086.2 | NP_001119558.1 | 2 - 3 | 156 | 121 | |
GenBank mRNA | BC166893.1 | - | 2 - 3 | 135 | 6919817 | 121 |
FM039224.1 | - | 2 - 3 | 119 | 121 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |