Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fshr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199237.1 | NP_954707.1 | 1 - 2 | 218 | 85 | |
GenBank mRNA | L02842.1 | - | 1 - 2 | 218 | 85 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 G-protein coupled receptor Rattus norvegicus Probe spans exons |