Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mfsd2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002729615.3 | XP_002729661.1 | 1 - 2 | 407 | 63 | |
XM_006239869.2 | XP_006239931.1 | 1 - 2 | 407 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Rattus norvegicus Probe spans exons |