Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr35l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001099340.1 | NP_001092810.1 | 7 - 8 | 837 | 64 | |
XM_006239885.2 | XP_006239947.1 | 7 - 8 | 853 | 64 | ||
GenBank mRNA | EF613262.1 | - | 7 - 8 | 837 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Rattus norvegicus Probe spans exons |