Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snx6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108711.1 | NP_001102181.1 | 7 - 8 | 653 | 110 | |
GenBank mRNA | BC168856.1 | - | 6 - 7 | 614 | 7458822 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |