Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snapin | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001170576.1 | NP_001164047.1 | 2 - 3 | 222 | 66 | |
GenBank mRNA | DY315040.1 | - | 2 - 3 | 227 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Rattus norvegicus Probe spans exons |