Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lancl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191728.1 | NP_001178657.1 | 4 - 5 | 1105 | 86 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Rattus norvegicus 1700 Probe spans exons |