Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Blmh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034163.1 | NP_001029335.1 | 4 - 5 | 636 | 68 | |
GenBank mRNA | BC128701.1 | - | 4 - 5 | 521 | 7379958 | 68 |
CB772927.1 | - | 2 - 3 | 208 | 68 | ||
CK363809.1 | - | 4 - 5 | 563 | 68 | ||
D87336.1 | - | 4 - 5 | 539 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protease Mammalian Gene Collection Rattus norvegicus Probe spans exons |