Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Xlr4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006229523.2 | XP_006229585.1 | 3 - 4 | 680 | 116 | |
XM_006229524.2 | XP_006229586.1 | 3 - 4 | 695 | 116 | ||
XM_006229527.2 | XP_006229589.1 | 3 - 4 | 300 | 116 | ||
XM_006229528.2 | XP_006229590.1 | 3 - 4 | 662 | 116 | ||
XM_006229529.2 | XP_006229591.1 | 3 - 4 | 677 | 116 | ||
XM_219738.8 | XP_219738.6 | 2 - 3 | 839 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |