Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Xlr4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006229523.2 | XP_006229585.1 | 7 - 8 | 1099 | 71 | |
XM_006229524.2 | XP_006229586.1 | 7 - 8 | 1114 | 71 | ||
XM_006229527.2 | XP_006229589.1 | 7 - 8 | 719 | 71 | ||
XM_006229528.2 | XP_006229590.1 | 7 - 8 | 1081 | 71 | ||
XM_006229529.2 | XP_006229591.1 | 7 - 8 | 1096 | 71 | ||
XM_219738.8 | XP_219738.6 | 6 - 7 | 1258 | 71 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons |