Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tcp10b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107461.1 | NP_001100931.1 | 7 - 8 | 1076 | 88 | |
XM_006227927.2 | XP_006227989.1 | 11 - 12 | 1385 | 88 | ||
XM_006227928.2 | XP_006227990.1 | 10 - 11 | 1283 | 88 | ||
XM_006227930.2 | XP_006227992.1 | 9 - 10 | 1142 | 88 | ||
XM_008758757.1 | XP_008756979.1 | 8 - 9 | 1177 | 88 | ||
XM_008758758.1 | XP_008756980.1 | 7 - 8 | 1036 | 88 | ||
XM_008758759.1 | XP_008756981.1 | 10 - 11 | 1322 | 88 | ||
GenBank mRNA | BC166440.1 | - | 9 - 10 | 1073 | 7455355 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Probe spans exons |