Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tspyl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191618.2 | NP_001178547.1 | 3 - 4 | 1103 | 79 | |
XM_006257038.2 | XP_006257100.1 | 3 - 4 | 1254 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Rattus norvegicus Probe spans exons |