Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Podnl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_213845.7 | XP_213845.6 | 7 - 8 | 853 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus Probe spans exons |