Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tex14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008768160.1 | XP_008766382.1 | 3 - 4 | 362 | 68 | |
XM_008768161.1 | XP_008766383.1 | 3 - 4 | 601 | 68 | ||
XM_008768162.1 | XP_008766384.1 | 3 - 4 | 362 | 68 | ||
XM_008768163.1 | XP_008766385.1 | 3 - 4 | 362 | 68 | ||
XM_008768164.1 | XP_008766386.1 | 2 - 3 | 465 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus Probe spans exons |