Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nt5m | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105785.1 | NP_001099255.1 | 2 - 3 | 341 | 104 | |
XM_006246460.2 | XP_006246522.1 | 2 - 3 | 788 | 104 | ||
GenBank mRNA | BC167058.1 | - | 2 - 3 | 397 | 7310205 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |