Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpatch8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001062937.5 | XP_001062937.3 | 4 - 5 | 485 | 58 | |
XM_006247478.2 | XP_006247540.1 | 2 - 3 | 1193 | 58 | ||
XM_006247479.2 | XP_006247541.1 | 5 - 6 | 385 | 58 | ||
XM_008768226.1 | XP_008766448.1 | 2 - 3 | 1461 | 58 | ||
XM_008768227.1 | XP_008766449.1 | 4 - 5 | 2415 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 Rattus norvegicus 1700 Inventoried Probe spans exons |