Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc6a16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006229207.2 | XP_006229269.1 | 3 - 4 | 734 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Transporter Detect Genomic DNA Rattus norvegicus Probe spans exons |