Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ms4a4c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106339.1 | NP_001099809.1 | 3 - 4 | 309 | 76 | |
XM_006231087.2 | XP_006231149.1 | 4 - 5 | 478 | 76 | ||
XM_006231088.2 | XP_006231150.1 | 4 - 5 | 478 | 76 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Rattus norvegicus Probe spans exons |