Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ms4a4c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106339.1 | NP_001099809.1 | 4 - 5 | 435 | 119 | |
XM_006231087.2 | XP_006231149.1 | 5 - 6 | 604 | 119 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |