Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Krt86 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008810.1 | NP_001008810.1 | 7 - 8 | 1254 | 121 | |
GenBank mRNA | AB355641.1 | - | 7 - 8 | 1260 | 121 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |